Assay Details
| Gene Symbol: E2f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275880.2 | NP_001262809.1 | 6 - 7 | 3248 | 62 | |
| NM_001300525.1 | NP_001287454.1 | 6 - 7 | 2837 | 62 | ||
| NM_001300526.1 | NP_001287455.1 | 7 - 8 | 3256 | 62 | ||
| NM_079713.3 | NP_524437.2 | 5 - 6 | 3580 | 62 | ||
| NM_169961.3 | NP_732646.1 | 7 - 8 | 2817 | 62 | ||
| NM_169962.3 | NP_732647.1 | 5 - 6 | 3381 | 62 | ||
| GenBank mRNA | BT016096.1 | - | 7 - 8 | 2790 | 62 | |
More Information
Set Membership: |
|

E2f1
Genomic Map