Assay Details
| Gene Symbol: CASK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104392.3 | NP_001097862.1 | 10 - 11 | 2505 | 63 | |
| NM_001104393.3 | NP_001097863.1 | 10 - 11 | 1074 | 63 | ||
| NM_001170210.2 | NP_001163681.1 | 15 - 16 | 2670 | 63 | ||
| NM_001275882.1 | NP_001262811.1 | 14 - 15 | 2616 | 63 | ||
| NM_001382187.1 | NP_001369027.1 | 7 - 8 | 1815 | 63 | ||
| NM_079717.5 | NP_524441.2 | 14 - 15 | 2616 | 63 | ||
| NM_169970.3 | NP_732661.1 | 9 - 10 | 2600 | 63 | ||
| NM_169971.3 | NP_732662.2 | 11 - 12 | 2559 | 63 | ||
| GenBank mRNA | BT044550.1 | - | 10 - 11 | 2465 | 63 | |
| BT046172.1 | - | 14 - 15 | 2600 | 63 | ||
More Information
Set Membership: |
|

CASK
Genomic Map