Assay Details
| Gene Symbol: Nrx-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170216.3 | NP_001163687.1 | 12 - 13 | 4848 | 91 | |
| NM_001275908.2 | NP_001262837.1 | 11 - 12 | 5177 | 91 | ||
| NM_001275909.2 | NP_001262838.1 | 11 - 12 | 5186 | 91 | ||
| NM_001275912.2 | NP_001262841.1 | 11 - 12 | 5483 | 91 | ||
| NM_001275913.2 | NP_001262842.1 | 11 - 12 | 5186 | 91 | ||
| NM_001300538.1 | NP_001287467.1 | 11 - 12 | 5186 | 91 | ||
| NM_079725.5 | NP_524449.1 | 12 - 13 | 5277 | 91 | ||
| GenBank mRNA | BT023898.1 | - | 12 - 13 | 4824 | 91 | |
More Information
Set Membership: |
|

Nrx-1
Genomic Map