Assay Details
| Gene Symbol: Nrx-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170216.3 | NP_001163687.1 | 13 - 14 | 5180 | 86 | |
| NM_001275908.2 | NP_001262837.1 | 12 - 13 | 5509 | 86 | ||
| NM_001275909.2 | NP_001262838.1 | 12 - 13 | 5518 | 86 | ||
| NM_001275910.2 | NP_001262839.1 | 12 - 13 | 5473 | 86 | ||
| NM_001275911.2 | NP_001262840.1 | 12 - 13 | 5539 | 86 | ||
| NM_001275913.2 | NP_001262842.1 | 12 - 13 | 5518 | 86 | ||
| NM_001300538.1 | NP_001287467.1 | 12 - 13 | 5518 | 86 | ||
| NM_079725.5 | NP_524449.1 | 13 - 14 | 5609 | 86 | ||
| GenBank mRNA | BT023898.1 | - | 13 - 14 | 5156 | 86 | |
More Information
Set Membership: |
|

Nrx-1
Genomic Map