Assay Details
Gene Symbol: lmd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170044.2 | NP_732811.1 | 1 - 2 | 1776 | 66 | |
GenBank mRNA | AY032609.1 | - | 1 - 2 | 1762 | 66 | |
AY121651.1 | - | 1 - 2 | 1762 | 66 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
