Assay Details
| Gene Symbol: orb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275941.1 | NP_001262870.1 | 3 - 4 | 1145 | 69 | |
| NM_079736.3 | NP_524460.2 | 3 - 4 | 1145 | 69 | ||
| GenBank mRNA | BT133433.1 | - | 3 - 4 | 1138 | 69 | |
More Information
Set Membership: |
|

orb
Genomic Map