Assay Details
| Gene Symbol: orb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170228.1 | NP_001163699.1 | 3 - 4 | 850 | 76 | |
| NM_001275941.1 | NP_001262870.1 | 4 - 5 | 1626 | 76 | ||
| NM_079736.3 | NP_524460.2 | 4 - 5 | 1626 | 76 | ||
| NM_170062.2 | NP_732848.1 | 2 - 3 | 1015 | 76 | ||
| NM_170063.2 | NP_732849.2 | 2 - 3 | 1015 | 76 | ||
| GenBank mRNA | BT001315.1 | - | 3 - 4 | 851 | 76 | |
| BT133433.1 | - | 4 - 5 | 1619 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BT001315.1, NM_001170228.1 transcripts.
Set Membership: |
|

orb
Genomic Map