Assay Details
| Gene Symbol: orb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170228.1 | NP_001163699.1 | 6 - 7 | 1937 | 68 | |
| NM_001275941.1 | NP_001262870.1 | 7 - 8 | 2713 | 68 | ||
| NM_079736.3 | NP_524460.2 | 7 - 8 | 2713 | 68 | ||
| NM_170062.2 | NP_732848.1 | 5 - 6 | 2102 | 68 | ||
| NM_170063.2 | NP_732849.2 | 5 - 6 | 2102 | 68 | ||
| NM_170064.2 | NP_732850.1 | 2 - 3 | 808 | 68 | ||
| NM_170065.2 | NP_732851.2 | 2 - 3 | 808 | 68 | ||
| GenBank mRNA | BT001315.1 | - | 6 - 7 | 1938 | 68 | |
| BT133433.1 | - | 7 - 8 | 2706 | 68 | ||
More Information
Set Membership: |
|

orb
Genomic Map