Assay Details
| Gene Symbol: pnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079737.3 | NP_524461.2 | 2 - 3 | 1108 | 74 | |
| GenBank mRNA | BT058033.1 | - | 2 - 3 | 1104 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pnt| Gene Symbol: pnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079737.3 | NP_524461.2 | 2 - 3 | 1108 | 74 | |
| GenBank mRNA | BT058033.1 | - | 2 - 3 | 1104 | 74 | |
Set Membership: |
|