Assay Details
| Gene Symbol: pnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079737.3 | NP_524461.2 | 4 - 5 | 1328 | 68 | |
| NM_170651.4 | NP_732857.1 | 2 - 3 | 772 | 68 | ||
| GenBank mRNA | BT058033.1 | - | 4 - 5 | 1324 | 68 | |
| BT133301.1 | - | 3 - 4 | 730 | 68 | ||
More Information
Set Membership: |
|

pnt
Genomic Map