Assay Details
| Gene Symbol: pnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275943.1 | NP_001262872.1 | 2 - 3 | 2362 | 84 | |
| NM_079737.3 | NP_524461.2 | 7 - 8 | 2721 | 84 | ||
| NM_170070.2 | NP_732858.1 | 2 - 3 | 2356 | 84 | ||
| NM_170651.4 | NP_732857.1 | 5 - 6 | 2165 | 84 | ||
| GenBank mRNA | BT058033.1 | - | 7 - 8 | 2717 | 84 | |
| BT133301.1 | - | 6 - 7 | 2123 | 84 | ||
| X69166.1 | - | 2699 | 84 | |||
More Information
Set Membership: |
|

pnt
Genomic Map