Assay Details
| Gene Symbol: pnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275943.1 | NP_001262872.1 | 3 - 4 | 2522 | 109 | |
| NM_079737.3 | NP_524461.2 | 8 - 9 | 2881 | 109 | ||
| NM_170070.2 | NP_732858.1 | 3 - 4 | 2516 | 109 | ||
| NM_170651.4 | NP_732857.1 | 6 - 7 | 2325 | 109 | ||
| GenBank mRNA | BT058033.1 | - | 8 - 9 | 2877 | 109 | |
| BT133301.1 | - | 7 - 8 | 2283 | 109 | ||
| X69166.1 | - | 2859 | 109 | |||
More Information
Set Membership: |
|

pnt
Genomic Map