Assay Details
| Gene Symbol: eIF3d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079739.4 | NP_524463.2 | 3 - 4 | 1285 | 81 | |
| GenBank mRNA | BT023923.1 | - | 3 - 4 | 1278 | 81 | |
More Information
Set Membership: |
|

eIF3d1
Genomic Map