Assay Details
| Gene Symbol: aux | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275315.1 | NP_001262244.1 | 6 - 7 | 2396 | 78 | |
| NM_141181.3 | NP_649438.1 | 6 - 7 | 2420 | 78 | ||
| NM_164317.3 | NP_730785.1 | 10 - 11 | 2518 | 78 | ||
| GenBank mRNA | BT031027.2 | - | 6 - 7 | 2356 | 78 | |
More Information
Set Membership: |
|

aux
Genomic Map