Assay Details
| Gene Symbol: MP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144530.2 | NP_001138002.1 | 1 - 2 | 152 | 64 | |
| NM_001316492.1 | NP_001303421.1 | 1 - 2 | 152 | 64 | ||
| NM_141193.4 | NP_649450.3 | 1 - 2 | 152 | 64 | ||
| GenBank mRNA | AF145611.1 | - | 1 - 2 | 147 | 64 | |
More Information
Set Membership: |
|

MP1
Genomic Map