Assay Details
| Gene Symbol: MP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141193.4 | NP_649450.3 | 2 - 3 | 311 | 68 | |
| GenBank mRNA | AF145611.1 | - | 2 - 3 | 306 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MP1| Gene Symbol: MP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141193.4 | NP_649450.3 | 2 - 3 | 311 | 68 | |
| GenBank mRNA | AF145611.1 | - | 2 - 3 | 306 | 68 | |
Set Membership: |
|