Assay Details
| Gene Symbol: Sfxn1-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141203.4 | NP_649460.3 | 3 - 4 | 726 | 63 | |
| NM_164345.3 | NP_730821.2 | 3 - 4 | 401 | 63 | ||
| NM_164346.3 | NP_730822.2 | 2 - 3 | 459 | 63 | ||
| NM_164347.3 | NP_730823.2 | 3 - 4 | 446 | 63 | ||
| GenBank mRNA | AY089419.1 | - | 3 - 4 | 382 | 63 | |
| BT001832.1 | - | 2 - 3 | 459 | 63 | ||
More Information
Set Membership: |
|

Sfxn1-3
Genomic Map