Assay Details
| Gene Symbol: Sfxn1-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141203.4 | NP_649460.3 | 6 - 7 | 1141 | 77 | |
| NM_164345.3 | NP_730821.2 | 6 - 7 | 816 | 77 | ||
| NM_164346.3 | NP_730822.2 | 5 - 6 | 874 | 77 | ||
| NM_164347.3 | NP_730823.2 | 6 - 7 | 861 | 77 | ||
| GenBank mRNA | AY089419.1 | - | 6 - 7 | 797 | 77 | |
| BT001832.1 | - | 5 - 6 | 874 | 77 | ||
More Information
Set Membership: |
|

Sfxn1-3
Genomic Map