Assay Details
| Gene Symbol: MED31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259979.2 | NP_001246908.1 | 3 - 4 | 281 | 71 | |
| NM_001275329.1 | NP_001262258.1 | 2 - 3 | 555 | 71 | ||
| NM_001300230.1 | NP_001287159.1 | 2 - 3 | 141 | 71 | ||
| NM_141226.4 | NP_649483.1 | 2 - 3 | 561 | 71 | ||
| NM_169013.3 | NP_730861.1 | 3 - 4 | 286 | 71 | ||
| GenBank mRNA | AY061423.1 | - | 2 - 3 | 539 | 71 | |
More Information
Set Membership: |
|

MED31
Genomic Map