Assay Details
| Gene Symbol: atms | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275334.1 | NP_001262263.1 | 4 - 5 | 453 | 65 | |
| NM_141236.3 | NP_649493.1 | 1 - 2 | 182 | 65 | ||
| GenBank mRNA | AY070561.1 | - | 1 - 2 | 166 | 65 | |
More Information
Set Membership: |
|

atms
Genomic Map