Assay Details
| Gene Symbol: atms | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275334.1 | NP_001262263.1 | 5 - 6 | 1415 | 63 | |
| NM_141236.3 | NP_649493.1 | 2 - 3 | 1144 | 63 | ||
| GenBank mRNA | AY070561.1 | - | 2 - 3 | 1128 | 63 | |
More Information
Set Membership: |
|

atms
Genomic Map