Assay Details
| Gene Symbol: ctrip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202241.2 | NP_001189170.1 | 9 - 10 | 8957 | 64 | |
| NM_001202242.2 | NP_001189171.1 | 8 - 9 | 7736 | 64 | ||
| NM_001202243.3 | NP_001189172.2 | 8 - 9 | 7745 | 64 | ||
| NM_001202244.2 | NP_001189173.1 | 9 - 10 | 8327 | 64 | ||
| NM_001202245.3 | NP_001189174.1 | 9 - 10 | 9129 | 64 | ||
| NM_001202246.3 | NP_001189175.2 | 10 - 11 | 8440 | 64 | ||
| NM_001202247.2 | NP_001189176.1 | 8 - 9 | 7748 | 64 | ||
| NM_001300232.1 | NP_001287161.1 | 8 - 9 | 8963 | 64 | ||
| NM_001300233.1 | NP_001287162.1 | 8 - 9 | 8240 | 64 | ||
| NM_001300234.1 | NP_001287163.1 | 7 - 8 | 7661 | 64 | ||
| NM_141239.5 | NP_649496.3 | 9 - 10 | 8959 | 64 | ||
More Information
Set Membership: |
|

ctrip
Genomic Map