Assay Details
| Gene Symbol: ctrip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202241.2 | NP_001189170.1 | 10 - 11 | 9143 | 78 | |
| NM_001202242.2 | NP_001189171.1 | 9 - 10 | 7922 | 78 | ||
| NM_001202243.3 | NP_001189172.2 | 9 - 10 | 7931 | 78 | ||
| NM_001202244.2 | NP_001189173.1 | 10 - 11 | 8513 | 78 | ||
| NM_001202245.3 | NP_001189174.1 | 10 - 11 | 9315 | 78 | ||
| NM_001202246.3 | NP_001189175.2 | 11 - 12 | 8626 | 78 | ||
| NM_001202247.2 | NP_001189176.1 | 9 - 10 | 7934 | 78 | ||
| NM_001300232.1 | NP_001287161.1 | 9 - 10 | 9149 | 78 | ||
| NM_001300233.1 | NP_001287162.1 | 9 - 10 | 8426 | 78 | ||
| NM_001300234.1 | NP_001287163.1 | 8 - 9 | 7847 | 78 | ||
| NM_141239.5 | NP_649496.3 | 10 - 11 | 9145 | 78 | ||
More Information
Set Membership: |
|

ctrip
Genomic Map