Assay Details
| Gene Symbol: ctrip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202241.2 | NP_001189170.1 | 11 - 12 | 9343 | 70 | |
| NM_001202242.2 | NP_001189171.1 | 10 - 11 | 8122 | 70 | ||
| NM_001202243.3 | NP_001189172.2 | 10 - 11 | 8131 | 70 | ||
| NM_001202244.2 | NP_001189173.1 | 11 - 12 | 8713 | 70 | ||
| NM_001202245.3 | NP_001189174.1 | 11 - 12 | 9515 | 70 | ||
| NM_001202246.3 | NP_001189175.2 | 12 - 13 | 8826 | 70 | ||
| NM_001202247.2 | NP_001189176.1 | 10 - 11 | 8134 | 70 | ||
| NM_001300232.1 | NP_001287161.1 | 10 - 11 | 9349 | 70 | ||
| NM_001300233.1 | NP_001287162.1 | 10 - 11 | 8626 | 70 | ||
| NM_001300234.1 | NP_001287163.1 | 9 - 10 | 8047 | 70 | ||
| NM_141239.5 | NP_649496.3 | 11 - 12 | 9345 | 70 | ||
More Information
Set Membership: |
|

ctrip
Genomic Map