Assay Details
| Gene Symbol: ctrip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202241.2 | NP_001189170.1 | 3 - 4 | 1882 | 56 | |
| NM_001202245.3 | NP_001189174.1 | 3 - 4 | 2057 | 56 | ||
| NM_001300232.1 | NP_001287161.1 | 3 - 4 | 1975 | 56 | ||
| NM_141239.5 | NP_649496.3 | 3 - 4 | 1884 | 56 | ||
More Information
Set Membership: |
|

ctrip
Genomic Map