Assay Details
| Gene Symbol: hd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001316495.1 | NP_001303424.1 | 1 - 2 | 513 | 79 | |
| NM_141274.3 | NP_649531.1 | 1 - 2 | 185 | 79 | ||
| GenBank mRNA | AY121655.1 | - | 1 - 2 | 182 | 79 | |
More Information
Set Membership: |
|

hd
Genomic Map