Assay Details
| Gene Symbol: Atg17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141316.2 | NP_649573.2 | 3 - 4 | 3957 | 102 | |
| NM_206434.2 | NP_996156.1 | 2 - 3 | 3936 | 102 | ||
| GenBank mRNA | BT133015.1 | - | 2 - 3 | 3934 | 102 | |
More Information
Set Membership: |
|

Atg17
Genomic Map