Assay Details
| Gene Symbol: Hpr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141337.2 | NP_649594.1 | 4 - 5 | 1459 | 74 | |
| GenBank mRNA | AJ556821.1 | - | 4 - 5 | 1351 | 74 | |
| AY122188.1 | - | 4 - 5 | 1425 | 74 | ||
More Information
Set Membership: |
|

Hpr1
Genomic Map