Assay Details
Gene Symbol: wa-cup | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141464.2 | NP_649721.2 | 3 - 4 | 694 | 118 | |
GenBank mRNA | AY119089.1 | - | 3 - 4 | 694 | 118 |

More Information
Set Membership: |
![]() ![]() |
