Assay Details
| Gene Symbol: wa-cup | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141464.2 | NP_649721.2 | 5 - 6 | 1078 | 75 | |
| GenBank mRNA | AY119089.1 | - | 5 - 6 | 1078 | 75 | |
More Information
Set Membership: |
|

wa-cup
Genomic Map