Assay Details
| Gene Symbol: pyd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141475.3 | NP_649732.1 | 3 - 4 | 1077 | 88 | |
| GenBank mRNA | AF333187.1 | - | 3 - 4 | 1033 | 88 | |
More Information
Set Membership: |
|

pyd3
Genomic Map