Assay Details
| Gene Symbol: Taf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144551.2 | NP_001138023.1 | 1 - 2 | 185 | 93 | |
| NM_141491.3 | NP_649748.1 | 2 - 3 | 426 | 93 | ||
| GenBank mRNA | BT025189.1 | - | 2 - 3 | 426 | 93 | |
More Information
Set Membership: |
|

Taf7
Genomic Map