Assay Details
| Gene Symbol: PIG-H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275429.1 | NP_001262358.1 | 1 - 2 | 667 | 97 | |
| NM_141498.3 | NP_649755.1 | 1 - 2 | 478 | 97 | ||
| GenBank mRNA | BT022579.1 | - | 1 - 2 | 629 | 97 | |
More Information
Set Membership: |
|

PIG-H
Genomic Map