Assay Details
| Gene Symbol: PIG-H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275429.1 | NP_001262358.1 | 2 - 3 | 752 | 83 | |
| NM_141498.3 | NP_649755.1 | 2 - 3 | 563 | 83 | ||
| GenBank mRNA | BT022579.1 | - | 2 - 3 | 714 | 83 | |
More Information
Set Membership: |
|

PIG-H
Genomic Map