Assay Details
| Gene Symbol: Ada2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031980.2 | NP_001027151.1 | 3 - 4 | 1167 | 81 | |
| NM_001275438.1 | NP_001262367.1 | 3 - 4 | 1237 | 81 | ||
| NM_141516.3 | NP_649773.1 | 3 - 4 | 1237 | 81 | ||
| GenBank mRNA | AY058546.1 | - | 3 - 4 | 1230 | 81 | |
| AY142213.1 | - | 3 - 4 | 1160 | 81 | ||
More Information
Set Membership: |
|

Ada2b
Genomic Map