Assay Details
| Gene Symbol: Ctr1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141533.4 | NP_649790.1 | 1 - 2 | 191 | 58 | |
| GenBank mRNA | AY071033.1 | - | 1 - 2 | 186 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctr1B| Gene Symbol: Ctr1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141533.4 | NP_649790.1 | 1 - 2 | 191 | 58 | |
| GenBank mRNA | AY071033.1 | - | 1 - 2 | 186 | 58 | |
Set Membership: |
|