Assay Details
| Gene Symbol: Atg13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141539.4 | NP_649796.1 | 1 | 2791 | 117 | |
| GenBank mRNA | AY061109.1 | - | 1 | 2763 | 117 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atg13| Gene Symbol: Atg13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141539.4 | NP_649796.1 | 1 | 2791 | 117 | |
| GenBank mRNA | AY061109.1 | - | 1 | 2763 | 117 | |
Set Membership: |
|