Assay Details
| Gene Symbol: IscU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141583.4 | NP_649840.1 | 2 - 3 | 738 | 138 | |
| GenBank mRNA | AY060228.1 | - | 2 - 3 | 722 | 138 | |
More Information
Set Membership: |
|

IscU
Genomic Map