Assay Details
| Gene Symbol: hyx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170093.2 | NP_001163564.1 | 3 - 4 | 1727 | 67 | |
| NM_141606.4 | NP_649863.1 | 2 - 3 | 1469 | 67 | ||
| GenBank mRNA | AY061525.1 | - | 2 - 3 | 1447 | 67 | |
| DQ305403.1 | - | 2 - 3 | 1447 | 67 | ||
More Information
Set Membership: |
|

hyx
Genomic Map