Assay Details
| Gene Symbol: Kdm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275471.1 | NP_001262400.1 | 2 - 3 | 306 | 156 | |
| NM_001275472.1 | NP_001262401.1 | 2 - 3 | 343 | 156 | ||
| NM_001275473.1 | NP_001262402.1 | 2 - 3 | 410 | 156 | ||
| NM_141607.3 | NP_649864.2 | 2 - 3 | 358 | 156 | ||
More Information
Set Membership: |
|

Kdm2
Genomic Map