Assay Details
| Gene Symbol: ATP6AP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141619.3 | NP_649876.1 | 2 - 3 | 490 | 73 | |
| GenBank mRNA | AY051790.1 | - | 2 - 3 | 455 | 73 | |
More Information
Set Membership: |
|

ATP6AP2
Genomic Map