Assay Details
| Gene Symbol: CG8135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141633.2 | NP_649890.1 | 1 - 2 | 702 | 120 | |
| GenBank mRNA | BT133230.1 | - | 1 - 2 | 711 | 120 | |
| Gene Symbol: BBIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170097.2 | NP_001163568.1 | 1 - 2 | 702 | 120 | |
| GenBank mRNA | AY119474.1 | - | 1 - 2 | 711 | 120 | |
More Information
Set Membership: |
|

CG8135
Genomic Map