Assay Details
| Gene Symbol: Pnn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104257.2 | NP_001097727.1 | 1 - 2 | 247 | 62 | |
| NM_141677.3 | NP_649934.1 | 1 - 2 | 247 | 62 | ||
| GenBank mRNA | AY071261.1 | - | 1 - 2 | 224 | 62 | |
More Information
Set Membership: |
|

Pnn
Genomic Map