Assay Details
| Gene Symbol: Whamy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043234.2 | NP_001036699.1 | 2 - 3 | 879 | 77 | |
| NM_001275506.1 | NP_001262435.1 | 2 - 3 | 536 | 77 | ||
| NM_001275507.1 | NP_001262436.1 | 3 - 4 | 777 | 77 | ||
| NM_001275508.1 | NP_001262437.1 | 3 - 4 | 786 | 77 | ||
| NM_141687.1 | NP_649944.1 | 2 - 3 | 545 | 77 | ||
More Information
Set Membership: |
|

Whamy
Genomic Map