Assay Details
| Gene Symbol: Art4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275516.1 | NP_001262445.1 | 1 - 2 | 584 | 67 | |
| NM_141706.3 | NP_649963.1 | 1 - 2 | 584 | 67 | ||
| GenBank mRNA | BT126023.1 | - | 1 - 2 | 579 | 67 | |
More Information
Set Membership: |
|

Art4
Genomic Map