Assay Details
| Gene Symbol: gammaSnap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141723.2 | NP_649980.1 | 1 | 822 | 85 | |
| GenBank mRNA | BT132830.1 | - | 1 | 822 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gammaSnap2| Gene Symbol: gammaSnap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141723.2 | NP_649980.1 | 1 | 822 | 85 | |
| GenBank mRNA | BT132830.1 | - | 1 | 822 | 85 | |
Set Membership: |
|