Assay Details
| Gene Symbol: Nuak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260090.1 | NP_001247019.1 | 10 - 11 | 4574 | 78 | |
| NM_001260091.1 | NP_001247020.1 | 9 - 10 | 3345 | 78 | ||
| NM_001275528.1 | NP_001262457.1 | 10 - 11 | 4124 | 78 | ||
| NM_001275529.1 | NP_001262458.1 | 9 - 10 | 3833 | 78 | ||
| NM_001382098.1 | NP_001369002.1 | 9 - 10 | 3833 | 78 | ||
| NM_001382099.1 | NP_001369003.1 | 9 - 10 | 3833 | 78 | ||
| NM_001382100.1 | NP_001369004.1 | 9 - 10 | 3383 | 78 | ||
| NM_001382101.1 | NP_001369005.1 | 10 - 11 | 4574 | 78 | ||
| NM_001382102.1 | NP_001369006.1 | 9 - 10 | 3833 | 78 | ||
| NM_001382103.1 | NP_001369007.1 | 10 - 11 | 4574 | 78 | ||
| NM_001382104.1 | NP_001369008.1 | 10 - 11 | 4124 | 78 | ||
| NM_001382105.1 | NP_001369009.1 | 9 - 10 | 3383 | 78 | ||
| NM_141734.2 | NP_649991.2 | 10 - 11 | 4086 | 78 | ||
| NM_141736.3 | NP_649993.2 | 9 - 10 | 3345 | 78 | ||
| NM_169337.2 | NP_731469.2 | 10 - 11 | 4574 | 78 | ||
| NM_206469.3 | NP_996191.1 | 9 - 10 | 3833 | 78 | ||
| NM_206470.1 | NP_996192.1 | 10 - 11 | 4291 | 78 | ||
| GenBank mRNA | BT050462.1 | - | 9 - 10 | 3833 | 78 | |
More Information
Set Membership: |
|

Nuak1
Genomic Map