Assay Details
| Gene Symbol: Nuak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260090.1 | NP_001247019.1 | 12 - 13 | 4846 | 77 | |
| NM_001382098.1 | NP_001369002.1 | 11 - 12 | 4105 | 77 | ||
| NM_001382099.1 | NP_001369003.1 | 11 - 12 | 4105 | 77 | ||
| NM_001382100.1 | NP_001369004.1 | 11 - 12 | 3655 | 77 | ||
| NM_001382102.1 | NP_001369006.1 | 11 - 12 | 4105 | 77 | ||
| NM_001382104.1 | NP_001369008.1 | 12 - 13 | 4396 | 77 | ||
| NM_001382105.1 | NP_001369009.1 | 11 - 12 | 3655 | 77 | ||
| NM_141734.2 | NP_649991.2 | 12 - 13 | 4358 | 77 | ||
| NM_141736.3 | NP_649993.2 | 11 - 12 | 3617 | 77 | ||
| NM_169337.2 | NP_731469.2 | 12 - 13 | 4846 | 77 | ||
| NM_206469.3 | NP_996191.1 | 11 - 12 | 4105 | 77 | ||
| NM_206470.1 | NP_996192.1 | 12 - 13 | 4563 | 77 | ||
| GenBank mRNA | BT050462.1 | - | 11 - 12 | 4105 | 77 | |
More Information
Set Membership: |
|

Nuak1
Genomic Map