Assay Details
| Gene Symbol: Bruce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275531.2 | NP_001262460.1 | 18 - 19 | 7833 | 87 | |
| NM_001275533.2 | NP_001262462.1 | 17 - 18 | 7530 | 87 | ||
| NM_001275534.2 | NP_001262463.1 | 17 - 18 | 7533 | 87 | ||
| NM_141738.5 | NP_649995.2 | 17 - 18 | 7533 | 87 | ||
| GenBank mRNA | AF517634.1 | - | 17 - 18 | 7530 | 87 | |
More Information
Set Membership: |
|

Bruce
Genomic Map