Assay Details
| Gene Symbol: Bruce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275531.2 | NP_001262460.1 | 26 - 27 | 10626 | 86 | |
| NM_001275532.2 | NP_001262461.1 | 24 - 25 | 10254 | 86 | ||
| NM_001275533.2 | NP_001262462.1 | 25 - 26 | 10323 | 86 | ||
| NM_001275534.2 | NP_001262463.1 | 25 - 26 | 10326 | 86 | ||
| NM_141738.5 | NP_649995.2 | 25 - 26 | 10326 | 86 | ||
| GenBank mRNA | AF517634.1 | - | 25 - 26 | 10323 | 86 | |
More Information
Set Membership: |
|

Bruce
Genomic Map